2-Chlor-4-fluorphenylisothiocyanat

Lieferant: FLUOROCHEM

008760-5G 008760-1G
FLUO008760-5GEA 134 EUR
FLUO008760-5G FLUO008760-1G
2-Chlor-4-fluorphenylisothiocyanat
2-Chlor-4-fluorphenylisothiocyanat
Formel: C₇H₃ClFNS
CAS-Nummer: 362601-84-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR