3-Chlor-2-fluoranisol

Lieferant: FLUOROCHEM

Synonyme: 1-Chloro-2-fluoro-3-methoxybenzene

005612-1G 005612-5G
FLUO005612-1GEA 31.1 EUR
FLUO005612-1G FLUO005612-5G
3-Chlor-2-fluoranisol
3-Chlor-2-fluoranisol
Formel: C₇H₆ClFO
Molecular Weight: 160.58 g/mol
MDL: MFCD01631570
CAS-Nummer: 261762-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR