4-Chlor-3-fluoranisol

Lieferant: FLUOROCHEM

Synonyme: 1-Chloro-2-fluoro-4-methoxybenzene

004420-5G 004420-10G 004420-50G 004420-25G
FLUO004420-5GEA 69.8 EUR
FLUO004420-5G FLUO004420-10G FLUO004420-50G FLUO004420-25G
4-Chlor-3-fluoranisol
4-Chlor-3-fluoranisol
Formel: C₇H₆ClFO
Molecular Weight: 160,58 g/mol
Siedepunkt: 196 °C (1013 hPa)
Dichte: 1,26 g/cm³ (20 °C)
MDL: MFCD00042570
CAS-Nummer: 501-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR