2-Chlor-6-fluoranisol

Lieferant: FLUOROCHEM
Achtung

005611-1G 005611-5G
FLUO005611-1GEA 31.1 EUR
FLUO005611-1G FLUO005611-5G
2-Chlor-6-fluoranisol
2-Chlor-6-fluoranisol
Formel: C₇H₆ClFO
Molecular Weight: 160.58 g/mol
MDL: MFCD01631573
CAS-Nummer: 53145-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR