3-Chlor-5-fluoranisol
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
1-Chloro-3-fluoro-5-methoxybenzene
FL13258.POR
013258-25G
CHMPFL13258.POREA
0
EUR
CHMPFL13258.POR
FLUO013258-25G
3-Chlor-5-fluoranisol
3-Chlor-5-fluoranisol
Formel:
C₇H₆ClFO Molecular Weight: 160.58 g/mol |
MDL:
MFCD00070770 CAS-Nummer: 202925-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...