2-Chlor-5-fluoranisol

Lieferant: CHEMPUR

CHMPFL4419.25EA 0 EUR
CHMPFL4419.25 CHMPFL4419.5
2-Chlor-5-fluoranisol
2-Chlor-5-fluoranisol
Formel: C₇H₆ClFO
Molecular Weight: 160,58 g/mol
Siedepunkt: 195 °C (1013 hPa)
Dichte: 1,28 g/cm³ (20 °C)
MDL: MFCD00042573
CAS-Nummer: 450-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR