2-Chlor-5-fluorphenol

Lieferant: FLUOROCHEM

004457-25G 004457-100G
FLUO004457-25GEA 104 EUR
FLUO004457-25G FLUO004457-100G
2-Chlor-5-fluorphenol
2-Chlor-5-fluorphenol
Formel: C₆H₄ClFO
Molecular Weight: 146,55 g/mol
Siedepunkt: 183 °C (1013 hPa)
MDL: MFCD00042524
CAS-Nummer: 3827-49-4
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR