3-Chlor-4-fluorbenzolsulfonylchlorid
Lieferant: Thermo Scientific
Synonyme:
3-Chlor-4-fluorbenzolsulfochlorid, 3-Chlor-4-fluorbenzol-1-sulfonylchlorid, 3-Chlor-4-fluorbenzolsulfonsäurechlorid
BTB05535.10
BTB05535.50
MAYBBTB05535.10EA
0
EUR
MAYBBTB05535.10
MAYBBTB05535.50
3-Chlor-4-fluorbenzolsulfonylchlorid
3-Chlor-4-fluorbenzolsulfonylchlorid
Formel:
C₆H₃Cl₂FO₂S Molecular Weight: 229.06 g/mol Siedepunkt: 104…105 °C (1 mmHg) Dichte: 1.61 g/cm³ (20 °C) |
MDL:
MFCD00041444 CAS-Nummer: 91170-93-3 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...