5-Chlor-2-fluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

013075-5G 013075-25G
FLUO013075-5GEA 42.9 EUR
FLUO013075-5G FLUO013075-25G
5-Chlor-2-fluorbenzolsulfonylchlorid
5-Chlor-2-fluorbenzolsulfonylchlorid
Formel: C₆H₃Cl₂FO₂S
Schmelzpunkt: 41…44 °C
MDL: MFCD03094221
CAS-Nummer: 351003-49-1
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR