4-Chlor-2-fluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 4-Chlor-2-fluorbenzolsulfochlorid, 4-Chlor-2-fluorbenzolsulfonsäurechlorid, 4-Chlor-2-fluorbenzol-1-sulfonylchlorid

013080-5G 013080-25G
FLUO013080-5GEA 146 EUR
FLUO013080-5G FLUO013080-25G
4-Chlor-2-fluorbenzolsulfonylchlorid
4-Chlor-2-fluorbenzolsulfonylchlorid
Formel: C₆H₃Cl₂FO₂S
Molecular Weight: 229.06 g/mol
MDL: MFCD03094560
CAS-Nummer: 141337-26-0
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR