2-Chlor-5-iodbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

631736-25G 631736-5G
SIAL631736-25GEA 66.8 EUR
SIAL631736-25G SIAL631736-5G
2-Chlor-5-iodbenzoesäure, Sigma-Aldrich®
2-Chlor-5-iodbenzoesäure
Formel: C₇H₄ClIO₂
Molecular Weight: 282,46 g/mol
MDL: MFCD00079731
CAS-Nummer: 19094-56-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR