2-Chlor-1-iod-4-nitrobenzol

Lieferant: FLUOROCHEM

Synonyme: 3-Chloro-4-iodonitrobenzene

CFF960.1 005496-1G 005496-5G 005496-25G
CHMPCFF960.1EA 522 EUR
CHMPCFF960.1 FLUO005496-1G FLUO005496-5G FLUO005496-25G
2-Chlor-1-iod-4-nitrobenzol
2-Chlor-1-iod-4-nitrobenzol
Formel: C₆H₃ClINO₂
Molecular Weight: 283,45 g/mol
Schmelzpunkt: 103 °C
MDL: MFCD00024589
CAS-Nummer: 41252-96-4
EINECS: 255-278-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR