2,3-Dichlorphenylisocyanat

Lieferant: Thermo Scientific

Synonyme: Isocyanic acid 2,3-Dichlorophenyl ester

SB01777.1 SB01777.10
MAYBSB01777.1EA 0 EUR
MAYBSB01777.1 MAYBSB01777.10
2,3-Dichlorphenylisocyanat
2,3-Dichlorphenylisocyanat
Formel: C₇H₃Cl₂NO
Molecular Weight: 188,01 g/mol
Siedepunkt: 247 °C (1013 hPa)
Dichte: 1,42 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00002000
CAS-Nummer: 41195-90-8
EINECS: 255-253-1
UN: 2250
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR