2,6-Dichlorphenylisocyanat

Lieferant: Thermo Scientific

CD00847.1 CD00847.10 CD00847.50
MAYBCD00847.1EA 0 EUR
MAYBCD00847.1 MAYBCD00847.10 MAYBCD00847.50
2,6-Dichlorphenylisocyanat
2,6-Dichlorphenylisocyanat
Formel: C₇H₃Cl₂NO
Molecular Weight: 188,01 g/mol
Siedepunkt: 101 °C (5 mmHg)
Schmelzpunkt: 42…44 °C
MDL: MFCD00002003
CAS-Nummer: 39920-37-1
EINECS: 254-699-4
UN: 2250
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR