2,4-Dichlorphenylisocyanat 96%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,4-Dichlorophenylisocyantae
L11089.06
L11089.14
L11089.03
L11089.06EA
52.1
EUR
L11089.06
L11089.14
L11089.03
2,4-Dichlorphenylisocyanat 96%
2,4-Dichlorphenylisocyanat
Purity: 96%
CAS No.: 2612-57-9
Molecular Formula: C7H3Cl2NO
Formula Weight: 188.01
Odour: Irritating
Melting Point: 57-61°
Boiling Point: 80°/1mm
Beilstein Reference: 2090780
UN No.: 2250
MDL No.: MFCD00002001
Formel:
C₇H₃Cl₂NO Molecular Weight: 188,01 g/mol Siedepunkt: 80 °C (1 mmHg) Schmelzpunkt: 57…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002001 CAS-Nummer: 2612-57-9 EINECS: 220-040-4 UN: 2250 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...