2,3-Dichlorphenylisothiocyanat
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
1,2-Dichlor-3-isothiocyanatobenzol
003514-5G
003514-25G
FLUO003514-5GEA
45.9
EUR
FLUO003514-5G
FLUO003514-25G
2,3-Dichlorphenylisothiocyanat
2,3-Dichlorphenylisothiocyanat
Formel:
C₇H₃Cl₂NS Molecular Weight: 204,08 g/mol Siedepunkt: 256…258 °C (1013 hPa) Dichte: 1,434 g/cm³ (20 °C) |
MDL:
MFCD00041048 CAS-Nummer: 6590-97-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...