2,3,4-Trichlorphenylisothiocyanat
Lieferant: FLUOROCHEM
Synonyme:
1-Isothiocyanato-2,3,4-trichlorobenzene
009578-1G
009578-5G
FLUO009578-1GEA
115
EUR
FLUO009578-1G
FLUO009578-5G
2,3,4-Trichlorphenylisothiocyanat
2,3,4-Trichlorphenylisothiocyanat
Formel:
C₇H₂Cl₃NS Siedepunkt: 305 °C (1013 hPa) Schmelzpunkt: 68…71 °C |
MDL:
MFCD00041049 CAS-Nummer: 127142-69-2 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...