2,4,5-Trichlorphenylisothiocyanat
Lieferant: FLUOROCHEM
009579-1G
009579-5G
FLUO009579-1GEA
67.1
EUR
FLUO009579-1G
FLUO009579-5G
2,4,5-Trichlorphenylisothiocyanat
2,4,5-Trichlorphenylisothiocyanat
Formel:
C₇H₂Cl₃NS Molecular Weight: 238,52 g/mol Siedepunkt: 280 °C (1013 hPa) Schmelzpunkt: 47…49 °C |
MDL:
MFCD00041052 CAS-Nummer: 23165-46-0 EINECS: 245-471-5 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...