2,4,6-Trichlorphenylisothiocyanat
Lieferant: FLUOROCHEM
Synonyme:
1-Isothiocyanato-2,4,6-trichlorobenzene
009580-1G
009580-5G
009580-25G
FLUO009580-1GEA
31.1
EUR
FLUO009580-1G
FLUO009580-5G
FLUO009580-25G
2,4,6-Trichlorphenylisothiocyanat
2,4,6-Trichlorphenylisothiocyanat
Formel:
C₇H₂Cl₃NS Molecular Weight: 238,52 g/mol Siedepunkt: 305 °C (1013 hPa) Schmelzpunkt: 80…81 °C |
MDL:
MFCD00041053 CAS-Nummer: 22134-07-2 EINECS: 244-798-0 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...