2-Chlor-4-nitrophenylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: 1-Chlor-2-isothiocyanato-5-nitrobenzol

008770-5G 008770-1G
FLUO008770-5GEA 279 EUR
FLUO008770-5G FLUO008770-1G
2-Chlor-4-nitrophenylisothiocyanat
2-Chlor-4-nitrophenylisothiocyanat
Formel: C₇H₃ClN₂O₂S
Molecular Weight: 214,63 g/mol
Schmelzpunkt: 102…104 °C
MDL: MFCD00041244
CAS-Nummer: 23165-64-2
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR