4-Amino-2-chlorphenol 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Chlor-4-hydroxyanilin
B23420.03
B23420.06
B23420.14
B23420.03EA
58
EUR
B23420.03
B23420.06
B23420.14
4-Amino-2-chlorphenol 99%
4-Amino-2-chlorphenol
Purity: 99%
CAS No.: 3964-52-1
Molecular Formula: C6H6ClNO
Formula Weight: 143.57
Melting Point: 150-153°
MDL No.: MFCD00128891
Formel:
C₆H₆ClNO Schmelzpunkt: 150…153 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128891 CAS-Nummer: 3964-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...