1-Chlor-3,5-dimethoxybenzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Chlorresorcindimethylether

B24834.06 B24834.14 B24834.22
B24834.06EA 36 EUR
B24834.06 B24834.14 B24834.22
1-Chlor-3,5-dimethoxybenzol ≥98%
1-Chlor-3,5-dimethoxybenzol

Purity: 98% CAS No.: 7051-16-3 Molecular Formula: C8H9ClO2 Formula Weight: 172.61 Flash Point: 112°(233°F) Melting Point: 32-36° Boiling Point: 234° Beilstein Reference: 2327304 MDL No.: MFCD00008382

Formel: C₈H₉ClO₂
Molecular Weight: 172,61 g/mol
Siedepunkt: 234 °C
Schmelzpunkt: 32…36 °C
Flammpunkt: 112 °C (233 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008382
CAS-Nummer: 7051-16-3
EINECS: 230-330-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR