2-Chlorthioanisol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2-Chlorphenylmethylsulfid, 1-Chlor-2-(methylthio)benzol

003552-5G 003552.10G 003552-25G
FLUO003552-5GEA 39.9 EUR
FLUO003552-5G FLUO003552.10G FLUO003552-25G
2-Chlorthioanisol
2-Chlorthioanisol
Formel: C₇H₇ClS
Molecular Weight: 158.65 g/mol
MDL: MFCD00040124
CAS-Nummer: 17733-22-1
UN: 3334
ADR: 9 null

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR