3-Chlorthioanisol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
3-Chlorphenylmethylsulfid, 1-Chlor-3-(methylthio)benzol
003553-5G
003553-25G
FLUO003553-5GEA
42.9
EUR
FLUO003553-5G
FLUO003553-25G
3-Chlorthioanisol
3-Chlorthioanisol
Formel:
C₇H₇ClS Molecular Weight: 158,65 g/mol Siedepunkt: 44 °C (0,08 mmHg) Dichte: 1,206 g/cm³ (25 °C) |
MDL:
MFCD00040127 CAS-Nummer: 4867-37-2 UN: 3334 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...