2,3,4-Trichlorbenzolsulfonylchlorid ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2,3,4-Trichlorbenzol-1-sulfonylchlorid, 2,3,4-Trichlorbenzolsulfonsäurechlorid, 2,3,4-Trichlorbenzolsulfochlorid

A15865.03 A15865.06
A15865.03EA 34 EUR
A15865.03 A15865.06
2,3,4-Trichlorbenzolsulfonylchlorid ≥97%
2,3,4-Trichlorbenzolsulfonylchlorid

Purity: 97% CAS No.: 34732-09-7 Molecular Formula: C6H2Cl4O2S Formula Weight: 279.96 Melting Point: 61-65° Beilstein Reference: 2649436 UN No.: 3261 MDL No.: MFCD00024871

Formel: C₆H₂Cl₄O₂S
Molecular Weight: 279,96 g/mol
Schmelzpunkt: 61…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00024871
CAS-Nummer: 34732-09-7
EINECS: 252-174-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR