2,3,4-Trichlorbenzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 2,3,4-Trichlorbenzol-1-sulfonylchlorid, 2,3,4-Trichlorbenzolsulfonsäurechlorid, 2,3,4-Trichlorbenzolsulfochlorid

TL00422.1 TL00422EA TL00422.50
MAYBTL00422.1EA 0 EUR
MAYBTL00422.1 MAYBTL00422EA MAYBTL00422.50
2,3,4-Trichlorbenzolsulfonylchlorid
2,3,4-Trichlorbenzolsulfonylchlorid
Formel: C₆H₂Cl₄O₂S
Molecular Weight: 279,96 g/mol
Schmelzpunkt: 65…66 °C
MDL: MFCD00024871
CAS-Nummer: 34732-09-7
EINECS: 252-174-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR