2,3-Dichlorbenzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2,3-Dichlorbenzolsulfonsäurechlorid, 2,3-Dichlorbenzol-1-sulfonylchlorid, 2,3-Dichlorbenzolsulfochlorid
003580-5G
003580-25G
FLUO003580-5GEA
39.9
EUR
FLUO003580-5G
FLUO003580-25G
2,3-Dichlorbenzolsulfonylchlorid
2,3-Dichlorbenzolsulfonylchlorid
Formel:
C₆H₃Cl₃O₂S Molecular Weight: 245,51 g/mol Siedepunkt: 140…142 °C (2 mmHg) Schmelzpunkt: 61…63 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00051844 CAS-Nummer: 82417-45-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...