2,4,5-Trichlorbenzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 2,4,5-Trichlorbenzol-1-sulfonylchlorid, 2,4,5-Trichlorbenzolsulfonsäurechlorid, 2,4,5-Trichlorbenzolsulfochlorid

SB01733.1 SB01733.10 SB01733.50
MAYBSB01733.1EA 0 EUR
MAYBSB01733.1 MAYBSB01733.10 MAYBSB01733.50
2,4,5-Trichlorbenzolsulfonylchlorid
2,4,5-Trichlorbenzolsulfonylchlorid
Formel: C₆H₂Cl₄O₂S
Molecular Weight: 279,96 g/mol
Schmelzpunkt: 69 °C
Dichte: 1,88 g/cm³ (20 °C)
MDL: MFCD00007428
CAS-Nummer: 15945-07-0
EINECS: 240-079-0
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR