2,4-Dichlorbenzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2,4-Dichlorbenzol-1-sulfonylchlorid, 2,4-Dichlorbenzolsulfochlorid, 2,4-Dichlorbenzolsulfonsäurechlorid
003581-25G
003581-10G
003581-100G
FLUO003581-25GEA
101
EUR
FLUO003581-25G
FLUO003581-10G
FLUO003581-100G
2,4-Dichlorbenzolsulfonylchlorid
2,4-Dichlorbenzolsulfonylchlorid
Formel:
C₆H₃Cl₃O₂S Molecular Weight: 245,51 g/mol Schmelzpunkt: 56 °C |
MDL:
MFCD00052712 CAS-Nummer: 16271-33-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...