2,5-Dichlorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2,5-Dichlorbenzolsulfochlorid, 2,5-Dichlorbenzolsulfonsäurechlorid, 2,5-Dichlorbenzol-1-sulfonylchlorid

003582-25G 003582-100G
FLUO003582-25GEA 79.2 EUR
FLUO003582-25G FLUO003582-100G
2,5-Dichlorbenzolsulfonylchlorid
2,5-Dichlorbenzolsulfonylchlorid
Formel: C₆H₃Cl₃O₂S
Schmelzpunkt: 39 °C
MDL: MFCD00007429
CAS-Nummer: 5402-73-3
EINECS: 226-453-6
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR