3-Chlorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 3-Chlorbenzolsulfonsäurechlorid, 3-Chlorbenzol-1-sulfonylchlorid, 3-Chlorbenzolsulfochlorid

003575-5G 003575-25G
FLUO003575-5GEA 0 EUR
FLUO003575-5G FLUO003575-25G
3-Chlorbenzolsulfonylchlorid
3-Chlorbenzolsulfonylchlorid
Formel: C₆H₄Cl₂O₂S
Molecular Weight: 211,07 g/mol
MDL: MFCD00051697
CAS-Nummer: 2888-06-4
EINECS: 220-753-0
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR