Methyl-2-cyanbenzoat

Lieferant: Thermo Scientific

Synonyme: 2-Cyanbenzoesäuremethylester

TL00681.1 TL00681.10 TL00681.50
MAYBTL00681.1EA 0 EUR
MAYBTL00681.1 MAYBTL00681.10 MAYBTL00681.50
Methyl-2-cyanbenzoat
Methyl-2-cyanbenzoat
Formel: C₉H₇NO₂
Molecular Weight: 161.16 g/mol
MDL: MFCD00016376
CAS-Nummer: 6587-24-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR