Methyl-2-cyanbenzoat
Lieferant: FLUOROCHEM
Synonyme:
2-Cyanbenzoesäuremethylester
F018829-1G
018829-5G
FLUOF018829-1GEA
55.4
EUR
FLUOF018829-1G
FLUO018829-5G
Methyl-2-cyanbenzoat
Methyl-2-cyanbenzoat
Formel:
C₉H₇NO₂ Molecular Weight: 161.16 g/mol |
MDL:
MFCD00016376 CAS-Nummer: 6587-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...