4-Ethoxybenzonitril ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
p-Ethoxybenzonitrile
A19806.06
A19806.14
A19806.22
A19806.06EA
39.3
EUR
A19806.06
A19806.14
A19806.22
4-Ethoxybenzonitril ≥98%
4-Ethoxybenzonitril
Purity: 98%
CAS No.: 25117-74-2
Molecular Formula: C9H9NO
Formula Weight: 147.18
Melting Point: 60-63°
Boiling Point: 258°
UN No.: 3439
MDL No.: MFCD00001819
Formel:
C₉H₉NO Molecular Weight: 147,18 g/mol Siedepunkt: 258 °C Schmelzpunkt: 60…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001819 CAS-Nummer: 25117-74-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...