2-Cyanbenzolsulfonylchlorid
Lieferant: Thermo Scientific
Synonyme:
2-Cyanbenzol-1-sulfonylchlorid, 2-Cyanbenzolsulfonsäurechlorid, 2-Cyanbenzolsulfochlorid
TL00431.0001
TL00431DA
TL00431.10
MAYBTL00431.0001EA
0
EUR
MAYBTL00431.0001
MAYBTL00431DA
MAYBTL00431.10
2-Cyanbenzolsulfonylchlorid
2-Cyanbenzolsulfonylchlorid
Formel:
C₇H₄ClNO₂S Molecular Weight: 201.63 g/mol Schmelzpunkt: 67,5 |
MDL:
MFCD00052341 CAS-Nummer: 69360-26-5 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...