3-Cyanbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 3-Cyanbenzolsulfochlorid, 3-Cyanbenzol-1-sulfonylchlorid, 3-Cyanbenzolsulfonsäurechlorid

CFF408.5 CFF408.10 CFF408.25 017431-5G 017431-25G 017431-1G
CHMPCFF408.5EA 375 EUR
CHMPCFF408.5 CHMPCFF408.10 CHMPCFF408.25 FLUO017431-5G FLUO017431-25G FLUO017431-1G
3-Cyanbenzolsulfonylchlorid
3-Cyanbenzolsulfonylchlorid
Formel: C₇H₄ClNO₂S
Molecular Weight: 201,63 g/mol
MDL: MFCD00203480
CAS-Nummer: 56542-67-7
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR