4-Cyanbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 4-Cyanbenzol-1-sulfonylchlorid, 4-Cyanbenzolsulfonsäurechlorid, 4-Cyanbenzolsulfochlorid

CFF407.5 CFF407.10 CFF407.25 017430-5G 017430-1G
CHMPCFF407.5EA 375 EUR
CHMPCFF407.5 CHMPCFF407.10 CHMPCFF407.25 FLUO017430-5G FLUO017430-1G
4-Cyanbenzolsulfonylchlorid
4-Cyanbenzolsulfonylchlorid
Formel: C₇H₄ClNO₂S
Molecular Weight: 201,63 g/mol
MDL: MFCD00052337
CAS-Nummer: 49584-26-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR