Benzoylchlorid ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Benzoesäurechlorid
A14107.30
A14107.0B
A14107.30EA
29.4
EUR
A14107.30
A14107.0B
Benzoylchlorid ≥99%
Benzoylchlorid
Purity: 99+%
CAS No.: 98-88-4
Molecular Formula: C7H5ClO
Formula Weight: 140.57
Flash Point: 69°(156°F)
Physical Form: Liquid
Density: 1.211
Refractive Index: 1.5535
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Pungent
pH: 2
Melting Point: -1°
Boiling Point: 197-199°
Merck Reference: 14,1112
UN No.: 1736
MDL No.: MFCD00000653
Formel:
C₇H₅ClO Molecular Weight: 140,57 g/mol Siedepunkt: 197…199 °C Schmelzpunkt: –1 °C Dichte: 1,211 Flammpunkt: 69 °C (156 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000653 CAS-Nummer: 98-88-4 EINECS: 202-710-8 UN: 1736 ADR: 8,II Merck Index: 14,1112 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...