Anthranilamid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Aminobenzamid, Anthranilsäureamid, 2-Aminobenzoesäureamid
A14756.22
A14756.36
A14756.22EA
30.3
EUR
A14756.22
A14756.36
Anthranilamid ≥98%
Anthranilamid
Purity: 98+%
CAS No.: 88-68-6
Molecular Formula: C7H8N2O
Formula Weight: 136.15
Flash Point: 198°(388°F)
Melting Point: 110-114°
Boiling Point: 300°
Beilstein Reference: 508509
MDL No.: MFCD00007981
Formel:
C₇H₈N₂O Molecular Weight: 136,15 g/mol Siedepunkt: 300 °C Schmelzpunkt: 110…114 °C Flammpunkt: 198 °C (388 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007981 CAS-Nummer: 88-68-6 EINECS: 201-851-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...