3-Aminobenzamid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Aminobenzoesäureamid

A10793.06 A10793.14
A10793.06EA 54.9 EUR
A10793.06 A10793.14
3-Aminobenzamid 98%
3-Aminobenzamid

Purity: 98% CAS No.: 3544-24-9 Molecular Formula: C7H8N2O Formula Weight: 136.15 Melting Point: 111-115° Beilstein Reference: 2802373

Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Siedepunkt: 341 °C (1013 hPa)
Schmelzpunkt: 111…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007989
CAS-Nummer: 3544-24-9
EINECS: 222-586-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR