4-Aminobenzamid 98

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Aminobenzoesäureamid

A12594.14 A12594.22 A12594.36
A12594.14EA 28.9 EUR
A12594.14 A12594.22 A12594.36
4-Aminobenzamid 98
4-Aminobenzamid

Purity: 98+% CAS No.: 2835-68-9 Molecular Formula: C7H8N2O Formula Weight: 136.15 Melting Point: 182-184° Beilstein Reference: 2079485

Formel: C₇H₈N₂O
Molecular Weight: 136.15 g/mol
Schmelzpunkt: 182…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007999
CAS-Nummer: 2835-68-9
EINECS: 220-612-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR