4-Aminobenzamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Aminobenzoesäureamid

M-Clarity™ Qualitätsstufe = MQ100

284572-100G 284572-25G
SIAL284572-100GEA 79.6 EUR
SIAL284572-100G SIAL284572-25G
4-Aminobenzamid, Sigma-Aldrich®
4-Aminobenzamid
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007999
CAS-Nummer: 2835-68-9
EINECS: 220-612-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR