Benzhydrazid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Benzoesäurehydrazid, Benzohydrazid

105771000 105775000
ACRO105771000EA 54 EUR
ACRO105771000 ACRO105775000
Benzhydrazid 98%
Benzhydrazid
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Schmelzpunkt: 113…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007596
CAS-Nummer: 613-94-5
EINECS: 210-363-9

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder or crystals or needles
Infrared spectrum Conforms
Melting point 113 to 117 °C
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR