Benzhydrazid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Benzoesäurehydrazid, Benzohydrazid
A12268.14
A12268.22
A12268.36
A12268.14EA
24.2
EUR
A12268.14
A12268.22
A12268.36
Benzhydrazid ≥98%
Benzhydrazid
Purity: 98%
CAS No.: 613-94-5
Molecular Formula: C7H8N2O
Formula Weight: 136.15
Melting Point: 113-117°
Beilstein Reference: 471797
Formel:
C₇H₈N₂O Molecular Weight: 136,15 g/mol Schmelzpunkt: 113…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007596 CAS-Nummer: 613-94-5 EINECS: 210-363-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...