Benzhydrazid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzoesäurehydrazid, Benzohydrazid

B13071-25G B13071-100G
SIALB13071-25GEA 50.3 EUR
SIALB13071-25G SIALB13071-100G
Benzhydrazid, Sigma-Aldrich®
Benzhydrazid
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Schmelzpunkt: 112…114 °C
MDL: MFCD00007596
CAS-Nummer: 613-94-5
EINECS: 210-363-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR