2-Nitrobenzhydrazid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Nitrobenzoic hydrazide

B21330.06 B21330.14
B21330.06EA 31.6 EUR
B21330.06 B21330.14
2-Nitrobenzhydrazid ≥98%
2-Nitrobenzhydrazid

Purity: 98% CAS No.: 606-26-8 Molecular Formula: C7H7N3O3 Formula Weight: 181.15 Melting Point: 119-123° Beilstein Reference: 394754

Formel: C₇H₇N₃O₃
Molecular Weight: 181,15 g/mol
Schmelzpunkt: 119…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007598
CAS-Nummer: 606-26-8
EINECS: 210-110-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR