o-Anisohydrazid

Lieferant: CHEMPUR

Synonyme: 2-Methoxybenzol-1-carbohydrazid, 2-Methoxybenzhydrazid, 2-Methoxybenzoesäurehydrazid, 2-Anisohydrazid, 2-Methoxybenzohydrazid

RH01067.250 RH01067.1
CHMPRH01067.250EA 0 EUR
CHMPRH01067.250 MAYBRH01067.1
o-Anisohydrazid
o-Anisohydrazid
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166.18 g/mol
MDL: MFCD00014755
CAS-Nummer: 7466-54-8
EINECS: 231-260-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR