m-Anisohydrazid ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
3-Methoxybenzoesäurehydrazid, 3-Methoxybenzhydrazid, 3-Methoxybenzol-1-carbohydrazid, 3-Anisohydrazid, 3-Methoxybenzohydrazid
B20956.06
B20956.14
B20956.06EA
25.1
EUR
B20956.06
B20956.14
m-Anisohydrazid ≥98%
m-Anisohydrazid
Purity: 98+%
CAS No.: 5785-06-8
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Melting Point: 90-94°
Beilstein Reference: 2092135
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166.18 g/mol Schmelzpunkt: 90…94 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00007601 CAS-Nummer: 5785-06-8 EINECS: 227-310-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...