Ethyl-2-isothiocyanatobenzoat

Lieferant: FLUOROCHEM

Synonyme: o-Ethoxycarbonylphenylisothiocyanat

008927-5G 008927-25G 008927-50G
FLUO008927-5GEA 134 EUR
FLUO008927-5G FLUO008927-25G FLUO008927-50G
Ethyl-2-isothiocyanatobenzoat
Ethyl-2-isothiocyanatobenzoat
Formel: C₁₀H₉NO₂S
Molecular Weight: 207.25 g/mol
Siedepunkt: 150…151 °C (1 mmHg)
Dichte: 1.201 g/cm³ (20 °C)
MDL: MFCD00041063
CAS-Nummer: 99960-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR