Methyl-2-isothiocyanatobenzoat
Lieferant: FLUOROCHEM
Synonyme:
o-Methoxycarbonylphenylisothiocyanat
008999-5G
008999-25G
FLUO008999-5GEA
137
EUR
FLUO008999-5G
FLUO008999-25G
Methyl-2-isothiocyanatobenzoat
Methyl-2-isothiocyanatobenzoat
Formel:
C₉H₇NO₂S Molecular Weight: 193,23 g/mol Siedepunkt: 104 °C (1013 hPa) Dichte: 1,25 g/cm³ (20 °C) |
MDL:
MFCD00041062 CAS-Nummer: 16024-82-1 EINECS: 240-164-2 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...